4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0%

Supplier: TCI America

M1637-1G M1637-5G
TCM1637-1GEA 317.42 CAD
TCM1637-1G TCM1637-5G
4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0%
4-Methoxyphenyl-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside

CAS Number: 1430068-18-0
MDL Number: MFCD06797146
Molecular Formula: C20H21N3O6
Molecular Weight: 399.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: -4.5 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₀H₂₁N₃O₆
MW: 399.40 g/mol
Melting Pt: 165 °C
MDL Number: MFCD06797146
CAS Number: 1430068-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0112687998a1b77eb0ce811d8b3add31752a89a2&vid=0112687998a1b77eb0ce811d8b3add31752a89a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR