4-Methoxyphenyl-2,6-di-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M1634-1G M1634-5G
TCM1634-1GEA 72.68 CAD
TCM1634-1G TCM1634-5G
4-Methoxyphenyl-2,6-di-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,6-di-O-benzyl-beta-D-galactopyranoside

CAS Number: 159922-50-6
MDL Number: MFCD06797145
Molecular Formula: C27H30O7
Molecular Weight: 466.53
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 113
Specific rotation [a]20/D: -1 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₇H₃₀O₇
Melting Pt: 113 °C
MDL Number: MFCD06797145
CAS Number: 159922-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9868bc9eed2a7ac976e4af966ad7524c3ad6db4&vid=c9868bc9eed2a7ac976e4af966ad7524c3ad6db4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR