4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M1633-1G
M1633-5G
TCM1633-1GEA
54.51
CAD
TCM1633-1G
TCM1633-5G
4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside
CAS Number: 159922-68-6
MDL Number: MFCD06797144
Molecular Formula: C30H34O7
Molecular Weight: 506.60
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 105
Specific rotation [a]20/D: 14 deg (C=1, CHCl3)
Storage Temperature: <0°C
Formula:
C₃₀H₃₄O₇ MW: 506.60 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD06797144 CAS Number: 159922-68-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...