4-Methoxyphenyl-β-D-glucopyranoside ≥97.0% (by HPLC)
Supplier: TCI America
M1631-5G
M1631-25G
TCM1631-5GEA
310.79
CAD
TCM1631-5G
TCM1631-25G
4-Methoxyphenyl-β-D-glucopyranoside ≥97.0% (by HPLC)
4-Methoxyphenyl-beta-D-glucopyranoside
CAS Number: 6032-32-2
MDL Number: MFCD06797143
Molecular Formula: C13H18O7
Molecular Weight: 286.28
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 176
Specific rotation [a]20/D: -64 deg (C=1, H2O)
Storage Temperature: <0°C
Formula:
C₁₃H₁₈O₇ Melting Pt: 176 °C |
MDL Number:
MFCD06797143 CAS Number: 6032-32-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...