Methyl-2,3,4-tri-O-acetyl-1-thio-β-L-fucopyranoside ≥98.0%

Supplier: TCI America

M1626-5G M1626-1G
TCM1626-5GEA 1143.76 CAD
TCM1626-5G TCM1626-1G
Methyl-2,3,4-tri-O-acetyl-1-thio-β-L-fucopyranoside ≥98.0%
Methyl-2,3,4-tri-O-acetyl-1-thio-beta-L-fucopyranoside

CAS Number: 84635-54-1
MDL Number: MFCD00080803
Molecular Formula: C13H20O7S
Molecular Weight: 320.36
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 148
Specific rotation [a]20/D: 1.5 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₁₃H₂₀O₇S
MW: 320.36 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00080803
CAS Number: 84635-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7531934c53abb7360aea1c46cd68eaaa54fcfea2&vid=7531934c53abb7360aea1c46cd68eaaa54fcfea2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR