4-Morpholinobenzaldehyde ≥98.0%

Supplier: TCI America

M1625-25G M1625-5G M1625-1G
TCM1625-25GEA 379.2 CAD
TCM1625-25G TCM1625-5G TCM1625-1G
4-Morpholinobenzaldehyde ≥98.0%
4-Morpholinobenzaldehyde

CAS Number: 1204-86-0
Molecular Formula: C11H13NO2
Molecular Weight: 191.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Other Product Names / Synonyms: 4-Morpholinobenzaldehyde

Formula: C₁₁H₁₃NO₂
Melting Pt: 63 °C
MDL Number: MFCD00735826
CAS Number: 1204-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=991d91963e2a5a3b91f01e6d8ed083b8ab1b7da4&vid=991d91963e2a5a3b91f01e6d8ed083b8ab1b7da4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR