4-(4-Methylpiperazin-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1624-25G M1624-5G
TCM1624-25GEA 366.88 CAD
TCM1624-25G TCM1624-5G
4-(4-Methylpiperazin-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)
4-(4-Methylpiperazin-1-yl)benzaldehyde

CAS Number: 27913-99-1
Molecular Formula: C12H16N2O
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63


Other Product Names / Synonyms: 1-(4-Formylphenyl)-4-methylpiperazine

Formula: C₁₂H₁₆N₂O
MW: 204.2682 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00452233
CAS Number: 27913-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c595df5556cb6dcb6d3d141bef437680d2b2a401&vid=c595df5556cb6dcb6d3d141bef437680d2b2a401&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR