(R)-(+)-N-(1-Phenylethyl)phthalamic acid ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-N-(1-Phenylethyl)phthalamic acid, (R)-(+)-N-(alpha-Methylbenzyl)phthalamic acid, (R)-2-((1-Phenylethyl)carbamoyl)benzoic acid

M1622-5G M1622-1G
TCM1622-5GEA 221.52 CAD
TCM1622-5G TCM1622-1G
(R)-(+)-N-(1-Phenylethyl)phthalamic acid ≥98.0%
(R)-(+)-N-(1-Phenylethyl)phthalamic acid

CAS Number: 21752-35-2
MDL Number: MFCD00063050
Molecular Formula: C16H15NO3
Molecular Weight: 269.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 128
Specific rotation [a]20/D: 46 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-N-(1-Phenylethyl)phthalamic Acid

Formula: C₁₆H₁₅NO₃
MW: 269.3
Melting Pt: 128 °C
MDL Number: MFCD00063050
CAS Number: 21752-35-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72af5f3c58e29452492f31808f01a724cef0d4a9&vid=72af5f3c58e29452492f31808f01a724cef0d4a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR