4-Methylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1621-25G
M1621-5G
TCM1621-25GEA
538.78
CAD
TCM1621-25G
TCM1621-5G
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
4-Methylcyclohexanecarboxylic acid (cis and trans mixture)
CAS Number: 4331-54-8
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 113
Specific Gravity (20/20): 1.01
Formula:
C₈H₁₄O₂ Boiling Pt: 140 °C Density: 1.01 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00074909 CAS Number: 4331-54-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...