4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥97.0% (by HPLC)

Supplier: TCI America

M1598-5G M1598-1G
TCM1598-5GEA 421.54 CAD
TCM1598-5G TCM1598-1G
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥97.0% (by HPLC)
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside

CAS Number: 889453-84-3
Molecular Formula: C31H29NO8
Molecular Weight: 543.57
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: 33 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₃₁H₂₉NO₈
Melting Pt: 130 °C
CAS Number: 889453-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e4b426fa731830feb8b9513a8cb684379213f61&vid=6e4b426fa731830feb8b9513a8cb684379213f61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR