2-Amino-2'-chloro-5-nitrobenzophenone ≥98.0%

Supplier: TCI America

A1754-5G A1754-25G
TCA1754-5GEA 101.12 CAD
TCA1754-5G TCA1754-25G
2-Amino-2'-chloro-5-nitrobenzophenone ≥98.0%
2-Amino-2'-chloro-5-nitrobenzophenone

CAS Number: 2011-66-7
MDL Number: MFCD00792453
Molecular Formula: C13H9ClN2O3
Molecular Weight: 276.68
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 121

Formula: C₁₃H₉ClN₂O₃
Melting Pt: 121 °C
MDL Number: MFCD00792453
CAS Number: 2011-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a5c7e443d048e20ab8f9de3882e2ed4fb0d4bc&vid=f2a5c7e443d048e20ab8f9de3882e2ed4fb0d4bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR