4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M1589-1G
M1589-5G
TCM1589-1GEA
85.54
CAD
TCM1589-1G
TCM1589-5G
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-beta-D-galactopyranoside
CAS Number: 400091-05-6
MDL Number: MFCD06797132
Molecular Formula: C23H26O7
Molecular Weight: 414.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 228
Specific rotation [a]20/D: -47.5 deg (C=0.5, Pyridine)
Storage Temperature: <0°C
Formula:
C₂₃H₂₆O₇ Melting Pt: 228 °C |
MDL Number:
MFCD06797132 CAS Number: 400091-05-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...