Methyl-4-methylsalicylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-hydroxy-4-methylbenzoate

M1584-25G M1584-5G
TCM1584-25GEA 123.24 CAD
TCM1584-25G TCM1584-5G
Methyl-4-methylsalicylate ≥98.0%
Methyl 4-methylsalicylate

CAS Number: 4670-56-8
MDL Number: MFCD00020130
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 242
Freezing point (°C): 28


Other Product Names / Synonyms: Methyl m-Cresotate, Methyl 2-Hydroxy-4-methylbenzoate, 4-Methylsalicylic Acid Methyl Ester

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 242 °C
MDL Number: MFCD00020130
CAS Number: 4670-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=104f6e7bcef3f19f80f4e268a64955ff2b4742ca&vid=104f6e7bcef3f19f80f4e268a64955ff2b4742ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR