Methyl-2,3,4-tri-O-benzoyl-ɑ-D-glucopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M1487-5G M1487-1G
TCM1487-5GEA 1673.22 CAD
TCM1487-5G TCM1487-1G
Methyl-2,3,4-tri-O-benzoyl-ɑ-D-glucopyranoside ≥98.0% (by HPLC)
Methyl-2,3,4-tri-O-benzoyl-alpha-D-glucopyranoside

CAS Number: 34234-44-1
MDL Number: MFCD06797128
Molecular Formula: C28H26O9
Molecular Weight: 506.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 53.5 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₈H₂₆O₉
Melting Pt: 146 °C
MDL Number: MFCD06797128
CAS Number: 34234-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fffc7a5535f237d9daaae928864ce05d8a38818&vid=2fffc7a5535f237d9daaae928864ce05d8a38818&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR