2-Methyl-1H-indole-3-carbaldehyde ≥98.0%

Supplier: TCI America

M1485-25G M1485-5G
TCM1485-25GEA 251.22 CAD
TCM1485-25G TCM1485-5G
2-Methyl-1H-indole-3-carbaldehyde ≥98.0%
2-Methyl-1H-indole-3-carbaldehyde

CAS Number: 5416-80-8
MDL Number: MFCD00012077
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 203


Other Product Names / Synonyms: 3-Formyl-2-methylindole

Formula: C₁₀H₉NO
Melting Pt: 203 °C
MDL Number: MFCD00012077
CAS Number: 5416-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af2b2708c15121ad901c4a4b91fdf914b34a1652&vid=af2b2708c15121ad901c4a4b91fdf914b34a1652&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR