4-Methoxyphenyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol

M1481-5G M1481-25G
TCM1481-5GEA 151.68 CAD
TCM1481-5G TCM1481-25G
4-Methoxyphenyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-beta-D-galactopyranoside

CAS Number: 3150-20-7
MDL Number: MFCD06797126
Molecular Formula: C13H18O7
Molecular Weight: 286.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 160
Specific rotation [a]20/D: -41 deg (C=1, MeOH)

Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Melting Pt: 160 °C
MDL Number: MFCD06797126
CAS Number: 3150-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d27b9c211bcfd92c8c8db949b4473f31f96e214&vid=1d27b9c211bcfd92c8c8db949b4473f31f96e214&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR