4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
M1477-5G
M1477-25G
TCM1477-5GEA
52.14
CAD
TCM1477-5G
TCM1477-25G
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-beta-D-galactopyranoside
CAS Number: 2872-65-3
MDL Number: MFCD06797123
Molecular Formula: C21H26O11
Molecular Weight: 454.43
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 105
Specific rotation [a]20/D: 3 deg (C=1, CHCl3)
Storage Temperature: <0°C
Formula:
C₂₁H₂₆O₁₁ MW: 454.43 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD06797123 CAS Number: 2872-65-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...