11-[4-(4-Butylphenylazo)phenoxy]undecyl methacrylate ≥97.0%
Supplier: TCI America
M1463-25G
M1463-5G
TCM1463-25GEA
1646.36
CAD
TCM1463-25G
TCM1463-5G
11-[4-(4-Butylphenylazo)phenoxy]undecyl methacrylate ≥97.0%
11-[4-(4-Butylphenylazo)phenoxy]undecyl methacrylate
CAS Number: 428515-74-6
MDL Number: MFCD06797121
Molecular Formula: C31H44N2O3
Molecular Weight: 492.70
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 76
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 11-[4-(4-Butylphenylazo)phenoxy]undecyl Ester
Formula:
C₃₁H₄₄N₂O₃ Melting Pt: 76 °C |
MDL Number:
MFCD06797121 CAS Number: 428515-74-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...