3-Methoxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1461-1G M1461-5G
TCM1461-1GEA 116.92 CAD
TCM1461-1G TCM1461-5G
3-Methoxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
3-Methoxy-2-naphthoic acid

CAS Number: 883-62-5
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134

Formula: C₁₂H₁₀O₃
Melting Pt: 134 °C
MDL Number: MFCD00094168
CAS Number: 883-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbdad79b2e2a506b72509ccefda69d90992f7356&vid=bbdad79b2e2a506b72509ccefda69d90992f7356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR