Methyltri-n-octylammonium Hydrogen sulfate ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
N-Methyl-N,N-dioctyloctan-1-aminium hydrogensulfate, Tri-n-octylmethylammonium Hydrogen Sulphate, Tri-n-octylmethylammonium Hydrogen sulfate, N-Methyl-N,N-dioctyloctan-1-aminium hydrogensulphate
M1457-5G
M1457-1G
TCM1457-5GEA
348.71
CAD
TCM1457-5G
TCM1457-1G
Methyltri-n-octylammonium Hydrogen sulfate ≥97.0% (by total nitrogen and titration analysis)
Methyltri-n-octylammonium Hydrogen sulfate
CAS Number: 59158-14-4
Molecular Formula: C25H55NO4S
Molecular Weight: 465.78
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Other Product Names / Synonyms: Tri-n-octylmethylammonium Hydrogen Sulfate
Formula:
C₂₅H₅₅NO₄S MW: 465.78 g/mol |
MDL Number:
MFCD03095545 CAS Number: 59158-14-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...