Methyltributylphosphonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1455-25G M1455-5G
TCM1455-25GEA 143.78 CAD
TCM1455-25G TCM1455-5G
Methyltributylphosphonium iodide ≥98.0% (by titrimetric analysis)
Methyltributylphosphonium iodide

CAS Number: 1702-42-7
MDL Number: MFCD00031626
Molecular Formula: C13H30IP
Molecular Weight: 344.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Methyltributylphosphonium Iodide

Formula: C₁₃H₃₀IP
MDL Number: MFCD00031626
CAS Number: 1702-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64dd94314795ceda7750f10f631db5c764a72811&vid=64dd94314795ceda7750f10f631db5c764a72811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR