ɑ-Methylene-γ-valerolactone ≥97.0% stabilized

Supplier: TCI America

Synonyms: 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone

M1453-5G M1453-25G
TCM1453-5GEA 181.7 CAD
TCM1453-5G TCM1453-25G
ɑ-Methylene-γ-valerolactone ≥97.0% stabilized
alpha-Methylene-gamma-valerolactone

(stabilized with HQ)
CAS Number: 62873-16-9
MDL Number: MFCD06797118
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 135
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone

Formula: C₆H₈O₂
MW: 112.13 g/mol
Density: 1.05 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD06797118
CAS Number: 62873-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4e926c75aac8a915077f917ea79c32e7f534d25&vid=a4e926c75aac8a915077f917ea79c32e7f534d25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR