3-(Methoxymethoxy)benzaldehyde ≥95.0%

Supplier: TCI America

M1448-25G M1448-5G
TCM1448-25GEA 611.62 CAD
TCM1448-25G TCM1448-5G
3-(Methoxymethoxy)benzaldehyde ≥95.0%
3-(Methoxymethoxy)benzaldehyde

CAS Number: 13709-05-2
MDL Number: MFCD04038425
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.14

Formula: C₉H₁₀O₃
Density: 1.14 (20 °C)
MDL Number: MFCD04038425
CAS Number: 13709-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4f9c081df05d215f3726a30bdac3cb0c9be4fc8&vid=c4f9c081df05d215f3726a30bdac3cb0c9be4fc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR