2-Methoxyethyl methanesulfonate ≥97.0%

Supplier: TCI America

Synonyms: Methanesulphonic Acid 2-Methoxyethyl Ester, Methanesulfonic Acid 2-Methoxyethyl Ester

M1446-5G M1446-25G
TCM1446-5GEA 165.9 CAD
TCM1446-5G TCM1446-25G
2-Methoxyethyl methanesulfonate ≥97.0%
2-Methoxyethyl methanesulfonate

CAS Number: 16427-44-4
MDL Number: MFCD02656401
Molecular Formula: C4H10O4S
Molecular Weight: 154.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 1.25


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Methanesulfonic Acid 2-Methoxyethyl Ester

Formula: C₄H₁₀O₄S
MW: 154.18 g/mol
Boiling Pt: 126 °C
Density: 1.25 (20 °C)
MDL Number: MFCD02656401
CAS Number: 16427-44-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d6bbb29c9b67bd0ab18776cf46526a6565bb26c&vid=9d6bbb29c9b67bd0ab18776cf46526a6565bb26c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR