2-Methyl-6-nitrobenzoic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1439-1G M1439-25G M1439-5G
TCM1439-1GEA 90.06 CAD
TCM1439-1G TCM1439-25G TCM1439-5G
2-Methyl-6-nitrobenzoic anhydride ≥98.0% (by titrimetric analysis)
2-Methyl-6-nitrobenzoic anhydride

CAS Number: 434935-69-0
MDL Number: MFCD06797117
Molecular Formula: C16H12N2O7
Molecular Weight: 344.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 177


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: 6-Nitro-o-toluic Anhydride, MNBA

Formula: C₁₆H₁₂N₂O₇
Melting Pt: 177 °C
MDL Number: MFCD06797117
CAS Number: 434935-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b334bee0ba509371fe6c1e7f8913ec6b26bf4e&vid=28b334bee0ba509371fe6c1e7f8913ec6b26bf4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR