4-Methoxyphthalic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1432-5G M1432-1G
TCM1432-5GEA 338.12 CAD
TCM1432-5G TCM1432-1G
4-Methoxyphthalic acid ≥98.0% (by titrimetric analysis)
4-Methoxyphthalic acid

CAS Number: 1885-13-8
MDL Number: MFCD00094739
Molecular Formula: C9H8O5
Molecular Weight: 196.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₉H₈O₅
MDL Number: MFCD00094739
CAS Number: 1885-13-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fcd1e78ce6be017770ce96a383820f30cc7ccbb&vid=9fcd1e78ce6be017770ce96a383820f30cc7ccbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR