2-Methylbenzo[e]indene (mixture of 1H- and 3H-form) ≥97.0%

Supplier: TCI America

M1425-1G M1425-100MG
TCM1425-1GEA 1380.13 CAD
TCM1425-1G TCM1425-100MG
2-Methylbenzo[e]indene (mixture of 1H- and 3H-form) ≥97.0%
2-Methylbenzo[e]indene (mixture of 1H- and 3H-form)

MDL Number: MFCD03701103
Molecular Formula: C14H12
Molecular Weight: 180.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.08

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bfac5b89c192bfced60192318632be3d4db5c1c&vid=e850b9e980c36ba6e76283c8754c542c6a4aa154&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR