4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) ≥98.0%

Supplier: TCI America

Synonyms: 1-(Hydroxymethyl)-4-methylcyclohexane

M1412-25G M1412-5G
TCM1412-25GEA 567.22 CAD
TCM1412-25G TCM1412-5G
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) ≥98.0%
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture)

CAS Number: 34885-03-5
MDL Number: MFCD00152628
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 1-(Hydroxymethyl)-4-methylcyclohexane

Formula: C₈H₁₆O
Density: 0.91 (20 °C)
MDL Number: MFCD00152628
CAS Number: 34885-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0800cefe0b3ac811a2011dd59745279c0ae9ba48&vid=0800cefe0b3ac811a2011dd59745279c0ae9ba48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR