Sodium-2-methyl-2-propene-1-sulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M1408-25G M1408-500G
TCM1408-25GEA 66.36 CAD
TCM1408-25G TCM1408-500G
Sodium-2-methyl-2-propene-1-sulfonate ≥98.0% (by HPLC, titration analysis)
Sodium-2-methyl-2-propene-1-sulfonate

CAS Number: 1561-92-8
MDL Number: MFCD00065344
Molecular Formula: C4H8O3S
Molecular Weight: 158.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: Methallylsulfonic Acid Sodium Salt, Sodium Methallylsulfonate, 2-Methyl-2-propene-1-sulfonic Acid Sodium Salt

Formula: C₄H₈O₃S
MDL Number: MFCD00065344
CAS Number: 1561-92-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=800d9c8e2c37a09316f55fdf746b4141369afcad&vid=800d9c8e2c37a09316f55fdf746b4141369afcad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR