5-Methyluridine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione
M1405-1G
M1405-25G
M1405-5G
TCM1405-1GEA
34.76
CAD
TCM1405-1G
TCM1405-25G
TCM1405-5G
5-Methyluridine ≥98.0% (by HPLC, titration analysis)
5-Methyluridine
CAS Number: 1463-10-1
MDL Number: MFCD00006535
Molecular Formula: C10H14N2O6
Molecular Weight: 258.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183
Specific rotation [a]20/D: -8 deg (C=2, H2O)
Formula:
C₁₀H₁₄N₂O₆ MW: 258.23 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD00006535 CAS Number: 1463-10-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...