(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M1402-500MG M1402-100MG
TCM1402-500MGEA 150.1 CAD
TCM1402-500MG TCM1402-100MG
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]

CAS Number: 175166-49-1
MDL Number: MFCD00799561
Molecular Formula: C21H18N2O2
Molecular Weight: 330.39
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 224
Specific rotation [a]20/D: -360 deg (C=1, CH2Cl2)

Formula: C₂₁H₁₈N₂O₂
Melting Pt: 224 °C
MDL Number: MFCD00799561
CAS Number: 175166-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abc88e8065b7e2854c51d7e27358753724432f19&vid=abc88e8065b7e2854c51d7e27358753724432f19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR