(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
[3aR-[2(3′aR*,8′aS*),3′aβ,8′aβ]]-(+)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole], Bis((3aR,8aS)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)methane, (+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
M1401-500MG
M1401-100MG
TCM1401-500MGEA
110.6
CAD
TCM1401-500MG
TCM1401-100MG
(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] ≥98.0% (by HPLC, total nitrogen)
(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
CAS Number: 180186-94-1
MDL Number: MFCD06797115
Molecular Formula: C21H18N2O2
Molecular Weight: 330.39
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 225
Specific rotation [a]20/D: 360 deg (C=1, CH2Cl2)
Formula:
C₂₁H₁₈N₂O₂ MW: 330.39 g/mol Melting Pt: 225 °C |
MDL Number:
MFCD06797115 CAS Number: 180186-94-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...