6-Methoxy-4-methylcoumarin ≥98.0%
Supplier: TCI America
M1398-25G
M1398-5G
TCM1398-25GEA
413.96
CAD
TCM1398-25G
TCM1398-5G
6-Methoxy-4-methylcoumarin ≥98.0%
6-Methoxy-4-methylcoumarin
CAS Number: 6295-35-8
MDL Number: MFCD00635101
Molecular Formula: C11H10O3
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164
Formula:
C₁₁H₁₀O₃ Melting Pt: 164 °C |
MDL Number:
MFCD00635101 CAS Number: 6295-35-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...