Minoxidil ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten

M1389-5G M1389-1G
TCM1389-5GEA 141.92 CAD
TCM1389-5G TCM1389-1G
Minoxidil ≥98.0%
Minoxidil

CAS Number: 38304-91-5
MDL Number: MFCD00063409
Molecular Formula: C9H15N5O
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Antihypertensive Ingredients, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2,6-Diamino-4-piperidinopyrimidine 1-Oxide

Formula: C₉H₁₅N₅O
MW: 209.253 g/mol
MDL Number: MFCD00063409
CAS Number: 38304-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e9a2532ee26e29423199440f1aaba2fc6e2ec57&vid=6e9a2532ee26e29423199440f1aaba2fc6e2ec57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR