2-(Methylamino)isobutyric acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,2-Dimethylalanine

M1388-5G M1388-1G
TCM1388-5GEA 304.94 CAD
TCM1388-5G TCM1388-1G
2-(Methylamino)isobutyric acid hydrate ≥98.0% (by titrimetric analysis)
2-(Methylamino)isobutyric acid hydrate

CAS Number: 2566-34-9
MDL Number: MFCD00004191
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: N,2-Dimethylalanine

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00004191
CAS Number: 2566-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4518a04568b41491ec714064413ad3d73cc6742b&vid=4518a04568b41491ec714064413ad3d73cc6742b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR