2-Amino-5-phenylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1684-1G A1684-5G
TCA1684-1GEA 130.51 CAD
TCA1684-1G TCA1684-5G
2-Amino-5-phenylpyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-phenylpyrazine

CAS Number: 13535-13-2
Molecular Formula: C10H9N3
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 146

Formula: C₁₀H₉N₃
Melting Pt: 146 °C
MDL Number: MFCD01646266
CAS Number: 13535-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4b56afb7e3ce0d636043c0dc057c3d2fe59fbb5&vid=c4b56afb7e3ce0d636043c0dc057c3d2fe59fbb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR