1,3-Benzodioxole-4-carbaldehyde ≥97.0%

Supplier: TCI America

Synonyms: Benzo[d][1,3]dioxole-4-carbaldehyde, 2,3-(Methylenedioxy)benzaldehyde, 1,3-Benzodioxole-4-carboxaldehyde

M1378-1G M1378-5G
TCM1378-1GEA 109.02 CAD
TCM1378-1G TCM1378-5G
1,3-Benzodioxole-4-carbaldehyde ≥97.0%
1,3-Benzodioxole-4-carbaldehyde

CAS Number: 7797-83-3
MDL Number: MFCD00016902
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 32


Other Product Names / Synonyms: 1,3-Benzodioxole-4-carboxaldehyde

Formula: C₈H₆O₃
Flash Pt: 110 °C
MDL Number: MFCD00016902
CAS Number: 7797-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=515c9d11280852bc0e822bc276a2f20fbfbac080&vid=515c9d11280852bc0e822bc276a2f20fbfbac080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR