2-(3-Methylphenyl)ethanol ≥98.0%
Supplier: TCI America
M1376-25G
M1376-5G
TCM1376-25GEA
556.16
CAD
TCM1376-25G
TCM1376-5G
2-(3-Methylphenyl)ethanol ≥98.0%
2-(3-Methylphenyl)ethanol
CAS Number: 1875-89-4
MDL Number: MFCD00002896
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Flash Point (°C): 109
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 3-Methylphenethyl Alcohol, 2-(3-Methylphenyl)ethyl Alcohol, 2-(3-Methylphenyl)ethanol
Formula:
C₉H₁₂O Boiling Pt: 243 °C Density: 1.00 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00002896 CAS Number: 1875-89-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...