Metrizoic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid

M1363-25G M1363-5G
TCM1363-025GEA 94.8 CAD
TCM1363-025G TCM1363-5G
Metrizoic acid ≥98.0%
Metrizoic acid

CAS Number: 1949-45-7
MDL Number: MFCD00867964
Molecular Formula: C12H11I3N2O4
Molecular Weight: 627.94
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal


Other Product Names / Synonyms: 3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzoic Acid

Formula: C₁₂H₁₁I₃N₂O₄
MW: 627.94 g/mol
MDL Number: MFCD00867964
CAS Number: 1949-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3fd42cabfe1653f0472acf874d7ffc0cb4a00d3&vid=b3fd42cabfe1653f0472acf874d7ffc0cb4a00d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR