N-Methyl-D-aspartic acid 98,0

Supplier: TCI America

Synonyms: N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid

M1360-100MG M1360-1G
TCM1360-100MGEA 36.34 CAD
TCM1360-100MG TCM1360-1G
N-Methyl-D-aspartic acid 98,0
N-Methyl-D-aspartic acid

CAS Number: 6384-92-5
MDL Number: MFCD00004226
Molecular Formula: C5H9NO4
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 178
Specific rotation [a]20/D: -15 deg (C=2, H2O)


Product Categories: Bioscience, Reagents for Other Disease Models, Biologically-modified Amino Acids
Other Product Names / Synonyms: N-Methyl-D-asparaginic Acid, Me-D-Asp-OH

Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00004226
CAS Number: 6384-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab55287b926d779e150b1d453a724da4464a7ade&vid=ab55287b926d779e150b1d453a724da4464a7ade&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR