Methyl-β-cyclodextrin (mixture of several Methylated)

Supplier: TCI America

Synonyms: Methylated-β-Cyclodextrin, Methyl-β-cyclodextrin

M1356-25G M1356-250G
TCM1356-025GEA 229.56 CAD
TCM1356-025G TCM1356-250G
Methyl-β-cyclodextrin (mixture of several Methylated)
Methyl-beta-cyclodextrin (mixture of several Methylated)

CAS Number: 128446-36-6
MDL Number: MFCD00074980
Form: Crystal
Color: White
Flash Point (°C): 187


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides, Cyclodextrins

Flash Pt: 187 °C
MDL Number: MFCD00074980
CAS Number: 128446-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ee3006962c4c01eead1cf63e4048d684955aa0a&vid=9ee3006962c4c01eead1cf63e4048d684955aa0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR