Benzyl-D-(-)-mandelate ≥98.0%
Supplier: TCI America
M1354-5G
M1354-25G
TCM1354-005GEA
56.25
CAD
TCM1354-005G
TCM1354-025G
Benzyl-D-(-)-mandelate ≥98.0%
Benzyl-D-(-)-mandelate
CAS Number: 97415-09-3
MDL Number: MFCD00674031
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107
Specific rotation [a]20/D: -34 deg (C=1, CH3CN)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Mandelic Acid Benzyl Ester
Formula:
C₁₅H₁₄O₃ Melting Pt: 107 °C |
MDL Number:
MFCD00674031 CAS Number: 97415-09-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...