Diethyl-D-(+)-malate ≥98.0%

Supplier: TCI America

Synonyms: D-(+)-Malic Acid Diethyl Ester, (R)-Diethyl 2-hydroxysuccinate, D-(+)-Apple Acid Diethyl Ester

M1351-5G M1351-25G
TCM1351-005GEA 204.93 CAD
TCM1351-005G TCM1351-25G
Diethyl-D-(+)-malate ≥98.0%
Diethyl-D-(+)-malate

CAS Number: 7554-28-1
MDL Number: MFCD02093483
Molecular Formula: C8H14O5
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.13


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester

Formula: C₈H₁₄O₅
MW: 190.20 g/mol
Density: 1.13 (20 °C)
MDL Number: MFCD02093483
CAS Number: 7554-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b84eb776c8ac198a01b1b730409dc2cc077caf1&vid=9b84eb776c8ac198a01b1b730409dc2cc077caf1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR