Diethyl-L-(-)-malate ≥97.0%

Supplier: TCI America

Synonyms: L-(-)-Malic Acid Diethyl Ester, L-(-)-Apple Acid Diethyl Ester

M1348-25G M1348-5G
TCM1348-25GEA 159.58 CAD
TCM1348-25G TCM1348-005G
Diethyl-L-(-)-malate ≥97.0%
Diethyl-L-(-)-malate

CAS Number: 691-84-9
MDL Number: MFCD00210119
Molecular Formula: C8H14O5
Molecular Weight: 190.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.13
Specific rotation [a]20/D: -12 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(-)-Apple Acid Diethyl Ester, L-(-)-Malic Acid Diethyl Ester

Formula: C₈H₁₄O₅
Density: 1.13 (20 °C)
MDL Number: MFCD00210119
CAS Number: 691-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=417aa48ac16266166903974bf0fda6c3b9b66535&vid=417aa48ac16266166903974bf0fda6c3b9b66535&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR