(+)-Dimethyl-D-malate ≥98.0%

Supplier: TCI America

Synonyms: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate

M1347-5G M1347-25G
TCM1347-005GEA 255.96 CAD
TCM1347-005G TCM1347-25G
(+)-Dimethyl-D-malate ≥98.0%
(+)-Dimethyl-D-malate

CAS Number: 70681-41-3
MDL Number: MFCD00066216
Molecular Formula: C6H10O5
Molecular Weight: 162.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.23
Specific rotation [a]20/D: 7 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester

Formula: C₆H₁₀O₅
MW: 162.1406 g/mol
Density: 1.23 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00066216
CAS Number: 70681-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf7c34028c7bd74a1ffa8424cad28c7894089e05&vid=bf7c34028c7bd74a1ffa8424cad28c7894089e05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR