Methyl nonafluorobutyl ether ≥98.0%

Supplier: TCI America

Synonyms: Perfluorobutyl Methyl Ether, Nonafluorobutyl Methyl Ether, Methyl Perfluorobutyl Ether

M1345-25G M1345-500G
TCM1345-025GEA 54.51 CAD
TCM1345-025G TCM1345-500G
Methyl nonafluorobutyl ether ≥98.0%
Methyl nonafluorobutyl ether

CAS Number: 163702-07-6
MDL Number: MFCD00674093
Molecular Formula: C5H3F9O
Molecular Weight: 250.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Methyl Perfluorobutyl Ether, Nonafluorobutyl Methyl Ether, Perfluorobutyl Methyl Ether

Formula: C₅H₃F₉O
MW: 250.06 g/mol
Density: 1.53 (20 °C)
MDL Number: MFCD00674093
CAS Number: 163702-07-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR