(R)-(-)-Ethyl mandelate ≥98.0%

Supplier: TCI America

Synonyms: (R)-(-)-Mandelic acid ethyl ester, Ethyl D-(-)-Mandelate, D(-)-Ethyl glycolate, D(-)-Mandelic acid ethyl ester, (R)-(-)-2-Hydroxy-2-phenylacetic acid ethyl ester, D(-)-Ethyl mandelate, (R)-(-)-Ethyl glycolate, (R)-Ethyl 2-hydroxy-2-phenylacetate, D(-)-2-Hydroxy-2-phenylacetic acid ethyl ester

M1344-25G M1344-5G
TCM1344-25GEA 388.36 CAD
TCM1344-25G TCM1344-005G
(R)-(-)-Ethyl mandelate ≥98.0%
(R)-(-)-Ethyl mandelate

CAS Number: 10606-72-1
MDL Number: MFCD00064248
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 29
Specific rotation [a]20/D: -138 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Mandelic Acid Ethyl Ester

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00064248
CAS Number: 10606-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=382e75b7e9d81b326eb1e08ed26aedec4502fcb6&vid=382e75b7e9d81b326eb1e08ed26aedec4502fcb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR