(-)-Dimethyl-L-malate ≥98.0%

Supplier: TCI America

M1343-25G M1343-5G
TCM1343-25GEA 82.16 CAD
TCM1343-25G TCM1343-005G
(-)-Dimethyl-L-malate ≥98.0%
(-)-Dimethyl-L-malate

CAS Number: 617-55-0
MDL Number: MFCD00066215
Molecular Formula: C6H10O5
Molecular Weight: 162.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 110
Specific Gravity (20/20): 1.23
Specific rotation [a]20/D: -7 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(-)-Apple Acid Dimethyl Ester, L-(-)-Malic Acid Dimethyl Ester

Formula: C₆H₁₀O₅
Boiling Pt: 108 °C
Density: 1.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00066215
CAS Number: 617-55-0
EINECS: 432-310-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0da672260fc8f3f9661be165babf42a47dac436d&vid=0da672260fc8f3f9661be165babf42a47dac436d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR