Furfuryl methacrylate ≥95.0% stabilized
Supplier: TCI America
M1335-5G
M1335-25G
TCM1335-005GEA
83.61
CAD
TCM1335-005G
TCM1335-025G
Furfuryl methacrylate ≥95.0% stabilized
Furfuryl methacrylate
(stabilized with MEHQ)
CAS Number: 3454-28-2
MDL Number: MFCD00046113
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 90
Specific Gravity (20/20): 1.09
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Furfuryl Ester
Formula:
C₉H₁₀O₃ Boiling Pt: 82 °C Density: 1.09 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00046113 CAS Number: 3454-28-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...