Methyl-2-methoxyphenylacetate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methoxyphenylacetic Acid Methyl Ester, Methyl 2-Methoxyphenylacetate

M1329-25G M1329-5G
TCM1329-025GEA 225.94 CAD
TCM1329-025G TCM1329-5G
Methyl-2-methoxyphenylacetate ≥98.0%
Methyl-2-methoxyphenylacetate

CAS Number: 27798-60-3
MDL Number: MFCD02093479
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2-Methoxyphenylacetic Acid Methyl Ester

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Density: 1.12 (20 °C)
MDL Number: MFCD02093479
CAS Number: 27798-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d039856902a3278fce801da624e54b81bcb7baf&vid=4d039856902a3278fce801da624e54b81bcb7baf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR